Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfonyl bromide, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
2297-65-6
C6H5BrO2S
221.07
CGWWQPZGKPHLBU-UHFFFAOYSA-N
phenylsulfonyl bromide
11138664
benzenesulfonyl bromide
C1=CC=C(C=C1)S(=O)(=O)Br
2297-65-6
C6H5BrO2S
221.07
CGWWQPZGKPHLBU-UHFFFAOYSA-N
phenylsulfonyl bromide
11138664
benzenesulfonyl bromide
C1=CC=C(C=C1)S(=O)(=O)Br
Boiling Point 83°C to 84°C (0.5 Torr)
Melting Point 19.5°C
Infrared Spectrum Authentic
Quantity 5g
Color Yellow to Brown
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 221.07
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Benzenesulfonyl bromide
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger