Alfa Aesar™ Benzenesulfonyl chloride, 98%

Manufacturer: Alfa Aesar™ A108490I

Quantity 5000g

 View more versions of this product

Catalog No. AAA108490I

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98-09-9
    Molecular Formula C6H5ClO2S
    Formula Weight 176.62
    MDL Number MFCD00007426
    UN Number UN2225
    Beilstein 606926
    Merck Index 14,1072
    Synonym Benzenesulfochloride
    Chemical Name or Material Benzenesulfonyl chloride

    Specifications

    Formula Weight 176.62
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DB8750000
    Melting Point 14° to 15°C
    Boiling Point 252°C
    MDL Number MFCD00007426
    Quantity 5000g
    CAS 98-09-9
    Beilstein 606926
    Odor Irritating
    Synonym Benzenesulfochloride
    TSCA TSCA
    Molecular Formula C6H5ClO2S
    Flash Point 132°C (270°F)
    UN Number UN2225
    Merck Index 14,1072
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BENZENE SULFONYL CHLORIDE
    Chemical Name or Material Benzenesulfonyl chloride
    Assay 98%

    Description

    Applications
    Benzenesulfonyl chloride reacts with Grignard reagents to form oxindoles from N-unsubstituted indoles. It is widely used to check the assay of thiamine in different food products. It is involved in the synthesis of alpha-disulfones, sulfonamides and sulfoante esters as precursor. It is a derivatization reagent for the determination of various amines in waste water and surface water at the sub-ppb level by gas chromatography-mass spectrometry. It is common reagent used in Hinsberg test for detection and distinguishing the type of amines as primary, secondary and tertiary amines.

    Solubility
    Insoluble in water. Soluble in ether and alcohol.

    Notes
    Moisture sensitive. Incompatible with water, strong oxidizing agents, strong bases, methyl formamide and dimethyl sulfoxide.

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit