Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfonyl Chloride 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
250mL
1L
5L
Packaging:
Glass bottle
Plastic drum
98-09-9
C6H5ClO2S
176.62
MFCD00007426
CSKNSYBAZOQPLR-UHFFFAOYSA-N
phenylsulfonyl chloride,benzene sulfochloride,benzenesulfonic chloride,benzenosulfochloride,benzene sulfonechloride,benzenosulfochlorek,benzenesulfonic acid chloride,benzenesulphonyl chloride,benzolsulfochloride,benzenesulfochloride
7369
benzenesulfonyl chloride
C1=CC=C(C=C1)S(=O)(=O)Cl
98-09-9
C6H5ClO2S
176.62
MFCD00007426
CSKNSYBAZOQPLR-UHFFFAOYSA-N
phenylsulfonyl chloride,benzene sulfochloride,benzenesulfonic chloride,benzenosulfochloride,benzene sulfonechloride,benzenosulfochlorek,benzenesulfonic acid chloride,benzenesulphonyl chloride,benzolsulfochloride,benzenesulfochloride
7369
benzenesulfonyl chloride
C1=CC=C(C=C1)S(=O)(=O)Cl
Specific Gravity 1.384
Color Undesignated
Boiling Point 251.0°C to 252.0°C
Melting Point 14.5°C
Flash Point 128°C
Density 1.3840g/mL
Quantity 5L
Packaging Plastic drum
Infrared Spectrum Authentic
Refractive Index 1.5508 to 1.5528
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5SO2Cl
Beilstein 11,34
Fieser 01,46; 05,22; 14,27
Merck Index 15,1073
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in alcohol and ether
Formula Weight 176.62
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Benzenesulfonyl chloride, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-636-6

RTECSNumber : DB8750000

TSCA : TSCA