Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfonyl fluoride, 98%, Thermo Scientific Chemicals

Catalog Number p-7044118
Click to view available options
Quantity:
1 g
5 g
368-43-4
C6H5FO2S
160.16
MFCD00007412
IDIPWEYIBKUDNY-UHFFFAOYSA-N
phenylsulfonyl fluoride, benzenesulphonyl fluoride, benzenesulfonylfluoride, phenylsulfonylfluoride, benzenesulphonylfluoride, fluorophenylsulfone, pubchem14507, acmc-20alq8, wln: wsfr, benzenesulfonyl fluoride
67779
benzenesulfonyl fluoride
FS(=O)(=O)C1=CC=CC=C1
368-43-4
C6H5FO2S
160.16
MFCD00007412
IDIPWEYIBKUDNY-UHFFFAOYSA-N
phenylsulfonyl fluoride, benzenesulphonyl fluoride, benzenesulfonylfluoride, phenylsulfonylfluoride, benzenesulphonylfluoride, fluorophenylsulfone, pubchem14507, acmc-20alq8, wln: wsfr, benzenesulfonyl fluoride
67779
benzenesulfonyl fluoride
FS(=O)(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.334
Boiling Point 203°C to 204°C
Flash Point 87°C (188°F)
Refractive Index 1.492
Quantity 1 g
UN Number UN2922
Beilstein 2206835
Sensitivity Moisture sensitive
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material Benzenesulfonyl fluoride
Show More Show Less
Hazard Category H227-H302+H312+H332-H314
Hazard Statement GHS H Statement
H301-H311-H331-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 206-707-2
RTECSNumber DB8940000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit