Alfa Aesar™ Benzenesulfonyl hydrazide, 98%

Manufacturer: Alfa Aesar™ B2397136

Quantity 500g

 View more versions of this product

Catalog No. AAB2397136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80-17-1
    Molecular Formula C6H8N2O2S
    Formula Weight 172.21
    MDL Number MFCD00007583
    UN Number UN3226
    Beilstein 640079
    Merck Index 14,7598
    Synonym Benzenesulfonohydrazide; BSH
    Chemical Name or Material Benzenesulfonyl hydrazide

    Specifications

    Formula Weight 172.21
    Molecular Formula C6H8N2O2S
    Melting Point 101° to 105°C
    MDL Number MFCD00007583
    Quantity 500g
    Merck Index 14,7598
    Health Hazard 3 GHS P Statement
    P210-P220-P301+P310-P405-P411a-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Store at temperatures not exceeding 194°F.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DB6888000
    TSCA TSCA
    CAS 80-17-1
    UN Number UN3226
    Beilstein 640079
    Synonym Benzenesulfonohydrazide; BSH
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H242-H228-H301
    Heating may cause a fire.
    Flammable solid.
    Toxic if swallowed.
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: SELF-REACTIVE SOLID TYPE D
    Chemical Name or Material Benzenesulfonyl hydrazide
    Assay 98%

    GHS H Statement
    H242-H228-H301
    Heating may cause a fire.
    Flammable solid.
    Toxic if swallowed.

    GHS P Statement
    P210-P220-P301+P310-P405-P411a-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Store at temperatures not exceeding 194°F.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit