Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzethonium Chloride, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
121-54-0
C27H42ClNO2
448.09
UREZNYTWGJKWBI-UHFFFAOYSA-M
benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
[Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1
121-54-0
C27H42ClNO2
448.09
UREZNYTWGJKWBI-UHFFFAOYSA-M
benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
[Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1

BE129, 121-54-0, C27H42ClNO2

Spectrum™ Benzethonium Chloride, USP exhibits a broad spectrum of microbiocidal activity against bacteria, fungi, mold and viruses. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Loss on Drying 5%
Packaging Fiber Drum
Quantity 50 kg
CAS Max % 100%
UN Number UN3261
Percent Purity 97 to 103%