Alfa Aesar™ Benzhydryl chloride, 98%

Manufacturer: Alfa Aesar™ B2476414

Quantity 25g

 View more versions of this product

Catalog No. AAB2476414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 90-99-3
    Molecular Formula C13H11Cl
    Formula Weight 202.69
    MDL Number MFCD00000855
    UN Number UN3265
    Beilstein 607925
    Synonym Chlorodiphenylmethane; Diphenylmethyl chloride
    Chemical Name or Material Benzhydryl chloride

    Specifications

    Formula Weight 202.69
    Boiling Point 139° to 140°C (3mmHg)
    Quantity 25g
    Odor Pungent
    TSCA TSCA
    Molecular Formula C13H11Cl
    MDL Number MFCD00000855
    CAS 90-99-3
    UN Number UN3265
    Synonym Chlorodiphenylmethane; Diphenylmethyl chloride
    Health Hazard 1 Danger
    Melting Point 17° to 20°C
    Flash Point 160°C (320°F)
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Beilstein 607925
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Benzhydryl chloride
    Assay 98%

    Description

    Applications
    Benzhydryl chloride was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.

    Solubility
    Insoluble in water.

    Notes
    Store in a tightly sealed container. Store away from oxidizing agents, bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit