Alfa Aesar™ Benzhydrylamine, 97%

Manufacturer: Alfa Aesar™ B2430306

Quantity 5g

 View more versions of this product

Catalog No. AAB2430306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 91-00-9
    Molecular Formula C13H13N
    Linear Formula (C6H5)2CHNH2
    Formula Weight 183.25
    MDL Number MFCD00008059
    UN Number UN2735
    Beilstein 776434
    Merck Index 14,1076
    Synonym Aminodiphenylmethane
    Chemical Name or Material Benzhydrylamine

    Specifications

    Formula Weight 183.25
    Molecular Formula C13H13N
    Linear Formula (C6H5)2CHNH2
    MDL Number MFCD00008059
    UN Number UN2735
    Merck Index 14,1076
    Synonym Aminodiphenylmethane
    Solubility Information Slightly soluble in water
    Melting Point 11° to 13°C
    Boiling Point 294° to 296°C
    Quantity 5g
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DA4407300
    CAS 91-00-9
    Beilstein 776434
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material Benzhydrylamine
    Assay 97%

    Description

    Applications
    Benzhydrylamine is useful for chiral resolution. It acts as an active pharmaceutical ingredient intermediate. It is also employed as a building block for chemical synthesis. Further, it is used in the preparation of furan-2-carboxylic acid benzhydrylamide by reacting with furan-2-carbonyl chloride.

    Solubility
    Miscible with chloroform, dimethylsulfoxide and methanol. Slightly miscible with water.

    Notes
    Air sensitive and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit