Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzhydrylamine, 97%

Click to view available options
Quantity:
5g
25g
100g
91-00-9
C13H13N
183.254
MFCD00008059
MGHPNCMVUAKAIE-UHFFFAOYSA-N
aminodiphenylmethane,benzhydrylamine,1,1-diphenylmethylamine,diphenylmethyl amine,1,1-diphenylmethanamine,alpha-aminodiphenylmethane,methanamine, 1,1-diphenyl,alpha-phenylbenzylamine,benzhydrylamin,benzhydryl amine
7036
diphenylmethanamine
C1=CC=C(C=C1)C(C2=CC=CC=C2)N
91-00-9
C13H13N
183.254
MFCD00008059
MGHPNCMVUAKAIE-UHFFFAOYSA-N
aminodiphenylmethane,benzhydrylamine,1,1-diphenylmethylamine,diphenylmethyl amine,1,1-diphenylmethanamine,alpha-aminodiphenylmethane,methanamine, 1,1-diphenyl,alpha-phenylbenzylamine,benzhydrylamin,benzhydryl amine
7036
diphenylmethanamine
C1=CC=C(C=C1)C(C2=CC=CC=C2)N
Applications
Benzhydrylamine is useful for chiral resolution. It acts as an active pharmaceutical ingredient intermediate. It is also employed as a building block for chemical synthesis. Further, it is used in the preparation of furan-2-carboxylic acid benzhydrylamide by reacting with furan-2-carbonyl chloride.

Solubility
Miscible with chloroform, dimethylsulfoxide and methanol. Slightly miscible with water.

Notes
Air sensitive and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
Boiling Point 294°C to 296°C
Quantity 100g
Melting Point 11°C to 13°C
Flash Point >110°C (230°F)
Linear Formula (C6H5)2CHNH2
UN Number UN2735
Beilstein 776434
Merck Index 14,1076
Solubility Information Slightly soluble in water
Formula Weight 183.25
Percent Purity 97%
Chemical Name or Material Benzhydrylamine
Show More Show Less

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : DA4407300