Alfa Aesar™ Benzhydrylidenefluorene, 98+%

Manufacturer: Alfa Aesar™ B2472214

Quantity 25g

 View more versions of this product

Catalog No. AAB2472214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4709-68-6
    Molecular Formula C26H18
    Formula Weight 330.43
    MDL Number MFCD00051452
    Beilstein 1977991
    Chemical Name or Material Benzhydrylidenefluorene

    Specifications

    Formula Weight 330.43
    Molecular Formula C26H18
    MDL Number MFCD00051452
    CAS 4709-68-6
    Beilstein 1977991
    Melting Point 230° to 232°C
    Quantity 25g
    Chemical Name or Material Benzhydrylidenefluorene
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit