Alfa Aesar™ Benzil dioxime, 98%

Manufacturer: Alfa Aesar™ A1068722

Quantity 100g

 View more versions of this product

Catalog No. AAA1068722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 23873-81-6
    Molecular Formula C14H12N2O2
    Formula Weight 240.26
    MDL Number MFCD00002113
    Beilstein 2053615
    Merck Index 14,1079
    Synonym Diphenylglyoxime
    Chemical Name or Material Benzil dioxime

    Specifications

    Formula Weight 240.26
    Molecular Formula C14H12N2O2
    CAS 23873-81-6
    Beilstein 2053615
    Merck Index 14,1079
    Synonym Diphenylglyoxime
    Melting Point ≈238°C (dec.)
    RTECS Number DD1985000
    TSCA TSCA
    MDL Number MFCD00002113
    Quantity 100g
    Chemical Name or Material Benzil dioxime
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit