Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzimidazole-6-thiocarboxamide, 97%

Click to view available options
Quantity:
250mg
1g
1520864-90-7
C8H7N3S
177.225
SWADWMCPVIOXDO-UHFFFAOYSA-N
benzimidazole-6-thiocarboxamide,1h-benzo d imidazole-6-carbothioamide,3h-1,3-benzodiazole-5-carbothioamide
82490593
3H-benzimidazole-5-carbothioamide
C1=CC2=C(C=C1C(=S)N)NC=N2
1520864-90-7
C8H7N3S
177.225
SWADWMCPVIOXDO-UHFFFAOYSA-N
benzimidazole-6-thiocarboxamide,1h-benzo d imidazole-6-carbothioamide,3h-1,3-benzodiazole-5-carbothioamide
82490593
3H-benzimidazole-5-carbothioamide
C1=CC2=C(C=C1C(=S)N)NC=N2
Color Brown to Yellow
Quantity 1g
Formula Weight 177.23
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Benzimidazole-6-thiocarboxamide