Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]furan-2-boronic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
98437-24-2
C8H7BO3
161.95
MFCD00236019
PKRRNTJIHGOMRC-UHFFFAOYSA-N
benzofuran-2-boronic acid,benzo b furan-2-boronic acid,benzofuran-2-ylboronic acid,2-benzofuranboronic acid,2-benzofuranylboronic acid,2,3-benzo b furan-2-boronic acid,2-boronobenzo b furan,1-benzofuran-2-yl boronic acid,1-benzofuran-2-boronic acid
2776266
1-benzofuran-2-ylboronic acid
B(C1=CC2=CC=CC=C2O1)(O)O
98437-24-2
C8H7BO3
161.95
MFCD00236019
PKRRNTJIHGOMRC-UHFFFAOYSA-N
benzofuran-2-boronic acid,benzo b furan-2-boronic acid,benzofuran-2-ylboronic acid,2-benzofuranboronic acid,2-benzofuranylboronic acid,2,3-benzo b furan-2-boronic acid,2-boronobenzo b furan,1-benzofuran-2-yl boronic acid,1-benzofuran-2-boronic acid
2776266
1-benzofuran-2-ylboronic acid
B(C1=CC2=CC=CC=C2O1)(O)O
Melting Point 112.0°C to 114.0°C
Quantity 5g
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: ca 2.5%
Formula Weight 161.95
Physical Form Powder
Percent Purity 97%
Chemical Name or Material Benzo[b]furan-2-boronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF