Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]furan-2-carboxylic acid, 98+%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
496-41-3
C9H6O3
162.144
MFCD00005848
OFFSPAZVIVZPHU-UHFFFAOYSA-N
benzofuran-2-carboxylic acid,coumarilic acid,benzo b furan-2-carboxylic acid,2-benzofurancarboxylic acid,2-carboxybenzofuran,coumarone-2-carboxylic acid,unii-7z6nmg947l,chembl84095,benzofuran-2-carboxylicacid
10331
1-benzofuran-2-carboxylic acid
C1=CC=C2C(=C1)C=C(O2)C(=O)O
496-41-3
C9H6O3
162.144
MFCD00005848
OFFSPAZVIVZPHU-UHFFFAOYSA-N
benzofuran-2-carboxylic acid,coumarilic acid,benzo b furan-2-carboxylic acid,2-benzofurancarboxylic acid,2-carboxybenzofuran,coumarone-2-carboxylic acid,unii-7z6nmg947l,chembl84095,benzofuran-2-carboxylicacid
10331
1-benzofuran-2-carboxylic acid
C1=CC=C2C(=C1)C=C(O2)C(=O)O
Boiling Point 310°C to 315°C
Melting Point 192°C to 196°C
Quantity 1g
Beilstein 124204
Merck Index 14,2561
Formula Weight 162.15
Percent Purity ≥98%
Chemical Name or Material Benzo[b]furan-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-818-9

RTECSNumber : DF6490000

TSCA : Yes

Recommended Storage : Ambient temperatures