Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]furan-3-boronic acid, 98%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
5g
317830-83-4
C8H7BO3
161.951
MFCD06801687
DFUGYZQSDFQVPU-UHFFFAOYSA-N
benzofuran-3-ylboronic acid,benzofuran-3-boronic acid,1-benzofuran-3-yl boronic acid,boronic acid, 3-benzofuranyl,acmc-1csxn,benzo b furan-3-boronic acid,benzofuran-3-yl boronic acid
23438904
1-benzofuran-3-ylboronic acid
B(C1=COC2=CC=CC=C12)(O)O
317830-83-4
C8H7BO3
161.951
MFCD06801687
DFUGYZQSDFQVPU-UHFFFAOYSA-N
benzofuran-3-ylboronic acid,benzofuran-3-boronic acid,1-benzofuran-3-yl boronic acid,boronic acid, 3-benzofuranyl,acmc-1csxn,benzo b furan-3-boronic acid,benzofuran-3-yl boronic acid
23438904
1-benzofuran-3-ylboronic acid
B(C1=COC2=CC=CC=C12)(O)O
Quantity 5g
Beilstein 9479377
Formula Weight 161.95
Physical Form Solid
Percent Purity 98%
Chemical Name or Material Benzo[b]furan-3-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C