Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]thiophene-2-carboxylic hydrazide, 99%, Acros Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
175135-07-6
C9H8N2OS
192.236
MFCD00052501
ZXKPFIRPUUAAPQ-UHFFFAOYSA-N
benzo b thiophene-2-carbohydrazide,benzo b thiophene-2-carboxylic hydrazide,thianaphthene-2-carboxylic hydrazide,benzo b thiophene-2-carboxylicacid, hydrazide,maybridge1_008989,acmc-1c3go,#,benzo b thiophene-2-carboxylic acid hydrazide
519437
1-benzothiophene-2-carbohydrazide
C1=CC=C2C(=C1)C=C(S2)C(=O)NN
175135-07-6
C9H8N2OS
192.236
MFCD00052501
ZXKPFIRPUUAAPQ-UHFFFAOYSA-N
benzo b thiophene-2-carbohydrazide,benzo b thiophene-2-carboxylic hydrazide,thianaphthene-2-carboxylic hydrazide,benzo b thiophene-2-carboxylicacid, hydrazide,maybridge1_008989,acmc-1c3go,#,benzo b thiophene-2-carboxylic acid hydrazide
519437
1-benzothiophene-2-carbohydrazide
C1=CC=C2C(=C1)C=C(S2)C(=O)NN
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 176.0°C to 178.0°C
Color White to Beige
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 192.24
Physical Form Needles
Percent Purity 99%
Chemical Name or Material Benzo[b]thiophene-2-carboxylic hydrazide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.