Alfa Aesar™ Benzo[b]thiophene-3-boronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H51876MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51876MD

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 171364-86-6
    Molecular Formula C14H17BO2S
    Formula Weight 260.16
    MDL Number MFCD11110552
    Synonym 2-Benzo[b]thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaboro lane; 4,4,5,5-Tetramethyl-2-benzo[b]thiophen-3-yl-1,3,2-dioxaborolane
    Chemical Name or Material Benzo[b]thiophene-3-boronic acid pinacol ester

    Specifications

    Formula Weight 260.16
    Molecular Formula C14H17BO2S
    MDL Number MFCD11110552
    CAS 171364-86-6
    Synonym 2-Benzo[b]thiophen-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaboro lane; 4,4,5,5-Tetramethyl-2-benzo[b]thiophen-3-yl-1,3,2-dioxaborolane
    Quantity 250mg
    Chemical Name or Material Benzo[b]thiophene-3-boronic acid pinacol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit