Alfa Aesar™ Benzo[c]cinnoline N-oxide, 95%

Manufacturer: Alfa Aesar™ B2254706

Quantity 5g

 View more versions of this product

Catalog No. AAB2254706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6141-98-6
    Molecular Formula C12H8N2O
    Formula Weight 196.21
    MDL Number MFCD00014475
    Beilstein 8732
    Chemical Name or Material Benzo[c]cinnoline N-oxide

    Specifications

    Formula Weight 196.21
    Molecular Formula C12H8N2O
    MDL Number MFCD00014475
    CAS 6141-98-6
    Beilstein 8732
    Melting Point 138° to 141°C
    Quantity 5g
    Odor Odorless
    RTECS Number DE7644000
    Chemical Name or Material Benzo[c]cinnoline N-oxide
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit