Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzo[c]cinnoline N-oxide, 95%

Manufacturer:  Alfa Aesar™ B2254703

Quantity 1g

 View more versions of this product

Catalog No. AAB2254703


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6141-98-6
196.21
8732
C12H8N2O
MFCD00014475
Benzo[c]cinnoline N-oxide

Specifications

6141-98-6
196.21
8732
1g
Benzo[c]cinnoline N-oxide
C12H8N2O
MFCD00014475
138° to 141°C
Odorless
95%

Safety and Handling

SDS & Safety Documents

RTECSNumber : DE7644000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit