Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[d]isoxazol-3-ol, 96%, Acros Organics

Click to view available options
Quantity:
1g
5g
21725-69-9
C7H5NO2
135.12
QLDQYRDCPNBPII-UHFFFAOYSA-N
benzo d isoxazol-3-ol,1,2-benzisoxazol-3 2h-one,1,2-benzoxazol-3-ol,1,2-benzisoxazol-3-ol,3-hydroxybenzisoxazole,3-hydroxy-1,2-benzisoxazole,1,2-benzisoxazolin-3-one,1,2-benzisoxazol-3-ol 8ci,2h-1,2-benzoxazol-3-one,benzisoxazolone
210830
1,2-benzoxazol-3-one
C1=CC=C2C(=C1)C(=O)NO2
21725-69-9
C7H5NO2
135.12
QLDQYRDCPNBPII-UHFFFAOYSA-N
benzo d isoxazol-3-ol,1,2-benzisoxazol-3 2h-one,1,2-benzoxazol-3-ol,1,2-benzisoxazol-3-ol,3-hydroxybenzisoxazole,3-hydroxy-1,2-benzisoxazole,1,2-benzisoxazolin-3-one,1,2-benzisoxazol-3-ol 8ci,2h-1,2-benzoxazol-3-one,benzisoxazolone
210830
1,2-benzoxazol-3-one
C1=CC=C2C(=C1)C(=O)NO2
Packaging Glass Bottle
Melting Point 137°C to 140°C
Quantity 1g
Assay Percent Range 95% min. (HPLC)
Formula Weight 135.12
Percent Purity 96%
Chemical Name or Material Benzo[d]isoxazol-3-ol

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF