Alfa Aesar™ Benzofurazan-5-boronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H29358MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH29358MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1073355-14-2
    Molecular Formula C12H15BN2O3
    Formula Weight 246.07
    MDL Number MFCD09842717
    Synonym 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furazan
    Chemical Name or Material Benzofurazan-5-boronic acid pinacol ester

    Specifications

    Formula Weight 246.07
    Molecular Formula C12H15BN2O3
    Quantity 250mg
    MDL Number MFCD09842717
    CAS 1073355-14-2
    Synonym 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furazan
    Melting Point 87° to 88°C
    Chemical Name or Material Benzofurazan-5-boronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit