Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Benzofuroxan, 98%
Click to view available options
Quantity:
10g
50g
Description
Applications
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 67°C to 71°C |
Quantity | 10g |
Beilstein | 125112 |
Solubility Information | Soluble in methanol. Insoluble in water. |
Formula Weight | 136.11 |
Percent Purity | 98% |
Chemical Name or Material | Benzofuroxan |
Safety and Handling
EINECSNumber : 207-559-1
TSCA : Yes
Recommended Storage : Ambient temperatures