Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoic acid 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
2.5kg
25kg
Packaging:
Fibre drum
Plastic bottle
65-85-0
C7H6O2
122.12
MFCD00002398
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv
243
CHEBI:30746
benzoic acid
C1=CC=C(C=C1)C(=O)O
65-85-0
C7H6O2
122.12
MFCD00002398
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzenecarboxylic acid,dracylic acid,benzeneformic acid,carboxybenzene,phenylformic acid,benzenemethanoic acid,phenylcarboxylic acid,retardex,benzoesaeure gk,benzoesaeure gv
243
CHEBI:30746
benzoic acid
C1=CC=C(C=C1)C(=O)O
Appearance (20 wt.% soln. in ethanol) clear
Boiling Point 249.0°C
Packaging Plastic bottle
Color Orange to White
Quantity 2.5kg
Infrared Spectrum Authentic
Water 0.5% Max. (K.F.)
Sulfated Ash 0.01 % max.
Melting Point 121.0°C to 123.0°C
Flash Point 121°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5CO2H
Beilstein 09,92
Fieser 01,49; 13,312
Merck Index 15,1093
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol,benzene,ether,chloroform,,ccl4,carbon disulfide and turpentine
Formula Weight 122.12
Physical Form Powder, Crystals and/or Chunks
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Benzoic acid
Show More Show Less

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger