Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoic Acid, Crystal, EP, BP, JP, USP, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
65-85-0
C7H6O2
122.12
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzoic acid
OC(=O)C1=CC=CC=C1
65-85-0
C7H6O2
122.12
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzoic acid
OC(=O)C1=CC=CC=C1

BE140, 65-85-0, C7H6O2

Spectrum™ Benzoic Acid, Crystal, USP, EP, BP, JP has antifungal and topical antiseptic properties. Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.05%
Loss on Drying 0.5%
Packaging Fiber Drum
Quantity 25 kg
Appearance Clear
CAS Max % 100%
Percent Purity 99.5 to 100.5%