Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzoic Acid, Crystal, Reagent, ACS, 99.5%, Spectrum™ Chemical
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Benzoic Acid, Crystal, Reagent, ACS an aromatic crystalline solid that is colorless. It is used as a standard for calibrating bomb calorimeters in teaching laboratories and as a synthetic precursor to a series of plasticizers. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
CAS Max % | 100% |
Packaging | Poly Pail |
Quantity | 2.5 kg |
Percent Purity | 99.5% |
Grade | ACS |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction