Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoic Acid, Crystal, Reagent, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
65-85-0
C7H6O2
122.12
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzoic acid
OC(=O)C1=CC=CC=C1
65-85-0
C7H6O2
122.12
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzoic acid
OC(=O)C1=CC=CC=C1

B1085, 65-85-0, C7H6O2

Spectrum™ Benzoic Acid, Crystal, Reagent, ACS an aromatic crystalline solid that is colorless. It is used as a standard for calibrating bomb calorimeters in teaching laboratories and as a synthetic precursor to a series of plasticizers. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Packaging Poly Pail
Quantity 12 kg
CAS Max % 100%
Percent Purity 99.5%