Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoic Acid, FCC, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
65-85-0
C7H6O2
122.12
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzoic acid
OC(=O)C1=CC=CC=C1
65-85-0
C7H6O2
122.12
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzoic acid
OC(=O)C1=CC=CC=C1

B1102, 65-85-0, C7H6O2

Spectrum™ Benzoic Acid, FCC is used as a food preservative inhibiting the growth of mold, yeast and some bacteria. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Water 0.7%
Ignition Residue 0.05%
Packaging Poly Pail
Quantity 2.5 kg
CAS Max % 100%
Percent Purity 99.5 to 100.5%