Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoic Acid, Sodium Salt, 99+%, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic Bottle
Plastic Bucket
532-32-1
C7H5NaO2
144.105
MFCD00012463
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate,benzoic acid, sodium salt,benzoic acid sodium salt,sobenate,antimol,benzoate sodium,benzoate of soda,benzoate, sodium,natrium benzoicum,caswell no. 746
517055
sodium;benzoate
C1=CC=C(C=C1)C(=O)[O-].[Na+]
532-32-1
C7H5NaO2
144.105
MFCD00012463
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate,benzoic acid, sodium salt,benzoic acid sodium salt,sobenate,antimol,benzoate sodium,benzoate of soda,benzoate, sodium,natrium benzoicum,caswell no. 746
517055
sodium;benzoate
C1=CC=C(C=C1)C(=O)[O-].[Na+]
CAS Min % 99.0
CAS Max % 100.0
Acidity Alkalinity 0.2mL 0.1M NaOH or HCl per gram max.
Assay Percent Range 99+%
Linear Formula C6H5CO2Na
Beilstein 09, 92
Fieser 01,1044; 02,377
Merck Index 15, 8718
Formula Weight 144.11
Physical Form Crystals, Granules, Flakes or Crystalline Powder
Percent Purity ≥99.0% (on dried substance)
Grade Extra Pure
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Packaging Plastic Bucket
Water 2% Max. (K.F.)
Color White
Quantity 5kg
Chemical Name or Material Sodium benzoate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.