Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoin, 99%, Thermo Scientific Chemicals

Catalog Number AAA1018830
Click to view available options
Quantity:
250 g
1000 g
5000 g
119-53-9
C14H12O2
212.25
MFCD00004496
ISAOCJYIOMOJEB-UHFFFAOYNA-N
benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone
8400
CHEBI:17682
2-hydroxy-1,2-diphenylethanone
OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
119-53-9
C14H12O2
212.25
MFCD00004496
ISAOCJYIOMOJEB-UHFFFAOYNA-N
benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone
8400
CHEBI:17682
2-hydroxy-1,2-diphenylethanone
OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.

Solubility
Soluble in chlorine. Slightly soluble in water, ethanol and ether.

Notes
Moisture sensitive. Incompatible with acids, acid anhydrides, acid chlorides and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 131°C to 136°C
Density 1.31
Boiling Point 334°C to 344°C
Flash Point 181°C (357°F)
Odor Pleasant
Quantity 250 g
Beilstein 391839
Merck Index 14,1093
Solubility Information Soluble in chlorine. Slightly soluble in water,ethanol and ether.
Formula Weight 212.25
Percent Purity 99%
Chemical Name or Material Benzoin
Show More Show Less
EINECSNumber 204-331-3
RTECSNumber DI1590000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit