Alfa Aesar™ Benzoin isopropyl ether, 98+%

Manufacturer: Alfa Aesar™ B2265422

Quantity 100g

 View more versions of this product

Catalog No. AAB2265422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6652-28-4
    Molecular Formula C17H18O2
    Formula Weight 254.33
    MDL Number MFCD00015025
    Beilstein 1977110
    Synonym alpha-Isopropoxy-alpha-phenylacetophenone
    Chemical Name or Material Benzoin isopropyl ether

    Specifications

    Formula Weight 254.33
    TSCA TSCA
    Molecular Formula C17H18O2
    MDL Number MFCD00015025
    CAS 6652-28-4
    Beilstein 1977110
    Synonym alpha-Isopropoxy-alpha-phenylacetophenone
    Boiling Point 180°C (10mmHg)
    Melting Point 76° to 79°C
    Quantity 100g
    Chemical Name or Material Benzoin isopropyl ether
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit