Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzoin isopropyl ether, 98+%

Click to view available options
Quantity:
25g
100g
500g
6652-28-4
C17H18O2
254.329
MFCD00015025
MSAHTMIQULFMRG-UHFFFAOYSA-N
benzoin isopropyl ether,2-isopropoxy-1,2-diphenylethanone,ethanone, 2-1-methylethoxy-1,2-diphenyl,2-isopropoxy-2-phenylacetophenone,benzoinisopropylether,acetophenone, 2-isopropoxy-2-phenyl,cbmicro_021426,2-1-methylethoxy-1,2-diphenylethan-1-one,cambridge id 5476007
110912
1,2-diphenyl-2-propan-2-yloxyethanone
CC(C)OC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2
6652-28-4
C17H18O2
254.329
MFCD00015025
MSAHTMIQULFMRG-UHFFFAOYSA-N
benzoin isopropyl ether,2-isopropoxy-1,2-diphenylethanone,ethanone, 2-1-methylethoxy-1,2-diphenyl,2-isopropoxy-2-phenylacetophenone,benzoinisopropylether,acetophenone, 2-isopropoxy-2-phenyl,cbmicro_021426,2-1-methylethoxy-1,2-diphenylethan-1-one,cambridge id 5476007
110912
1,2-diphenyl-2-propan-2-yloxyethanone
CC(C)OC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2
Boiling Point 180°C (10mmHg)
Quantity 500g
Melting Point 76°C to 79°C
Beilstein 1977110
Formula Weight 254.33
Percent Purity ≥98%
Chemical Name or Material Benzoin isopropyl ether

EINECSNumber : 229-677-2

TSCA : Yes

Recommended Storage : Ambient temperatures