Alfa Aesar™ Benzonitrile, 99%

Manufacturer: Alfa Aesar™ A1117336

Quantity 500g

 View more versions of this product

Catalog No. AAA1117336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-47-0
    Molecular Formula C7H5N
    Formula Weight 103.12
    MDL Number MFCD00001770
    UN Number UN2224
    Beilstein 506893
    Merck Index 14,1097
    Chemical Name or Material Benzonitrile

    Specifications

    Formula Weight 103.12
    Health Hazard 3 GHS P Statement
    P210-P280-P301+P312-P312-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H5N
    Merck Index 14,1097
    Quantity 500g
    RTECS Number DI2450000
    TSCA TSCA
    CAS 100-47-0
    UN Number UN2224
    Melting Point −13°C
    MDL Number MFCD00001770
    Beilstein 506893
    Boiling Point 191° to 193°C
    Health Hazard 1 Warning
    Flash Point 71°C (159°F)
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BENZONITRILE
    Health Hazard 2 GHS H Statement
    H302-H312-H227
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.
    Chemical Name or Material Benzonitrile
    Assay 99%

    Description

    Applications
    Benzonitrile is a widely utilized as a solvent and an intermediate in industries making drugs, perfumes, dyes, rubber, textiles, resins and specialty lacquers. It finds application as a versatile precursor for many derivatives. It coordinates with transition metal to form complexes which act as synthetic intermediates.

    Solubility
    Miscible with common organic solvents. Immiscible with water.

    Notes
    Hygroscopic. Incompatible with strong bases, strong oxidizing agents, reducing agents, acids, chlorates, nitrates and plastics.

    GHS H Statement
    H302-H312-H227
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.

    GHS P Statement
    P210-P280-P301+P312-P312-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit