Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzophenone imine, 97%, stabilized, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1013-88-3,123-31-9
C13H11N
181.23
MFCD00001760
SXZIXHOMFPUIRK-UHFFFAOYSA-N
benzophenone imine,benzophenoneimine,benzhydrylideneamine,1,1-diphenylmethanimine,unii-ejj21na7vi,benzhydrylimine,benzophenonimine,diphenylmethanimineimine,1,1-diphenylmethylimine,benzenemethanimine, .alpha.-phenyl
136809
diphenylmethanimine
C1=CC=C(C=C1)C(=N)C2=CC=CC=C2
1013-88-3,123-31-9
C13H11N
181.23
MFCD00001760
SXZIXHOMFPUIRK-UHFFFAOYSA-N
benzophenone imine,benzophenoneimine,benzhydrylideneamine,1,1-diphenylmethanimine,unii-ejj21na7vi,benzhydrylimine,benzophenonimine,diphenylmethanimineimine,1,1-diphenylmethylimine,benzenemethanimine, .alpha.-phenyl
136809
diphenylmethanimine
C1=CC=C(C=C1)C(=N)C2=CC=CC=C2
Melting Point -30°C
Boiling Point 216°C (75.0mmHg)
Flash Point 145°C
Specific Gravity 1.08
Viscosity 33,4 mPa.s (20°C)
Refractive Index 1.6170 to 1.619
Color Brown-Yellow to Colorless
Stabilizer approx. 0.02% w/w hydroquinone
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.0800g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (C6H5)2C=NH
Beilstein 07, IV, 1365
Merck Index 15, 1102
Solubility Information Solubility in water: 0.3g/L (25°C). Other solubilities: better soluble in aq. solution at lower ph values,,very soluble in diethyl ether
Formula Weight 181.23
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Benzophenone imine, Stabilized
Show More Show Less

GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning