Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzothiazole, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
250mL
1L
2.5L
Packaging:
Glass bottle
95-16-9
C7H5NS
135.18
MFCD00005775
IOJUPLGTWVMSFF-UHFFFAOYSA-N
benzothiazole,benzo d thiazole,benzosulfonazole,1-thia-3-azaindene,vangard bt,benzothiazol,usaf ek-4812,benzthiazole,fema number 3256,unii-g5bw2593ep
7222
CHEBI:45993
1,3-benzothiazole
C1=CC=C2C(=C1)N=CS2
95-16-9
C7H5NS
135.18
MFCD00005775
IOJUPLGTWVMSFF-UHFFFAOYSA-N
benzothiazole,benzo d thiazole,benzosulfonazole,1-thia-3-azaindene,vangard bt,benzothiazol,usaf ek-4812,benzthiazole,fema number 3256,unii-g5bw2593ep
7222
CHEBI:45993
1,3-benzothiazole
C1=CC=C2C(=C1)N=CS2
Melting Point 2°C
Flash Point >112°C
Boiling Point 231°C
Density 1.2300g/mL
Quantity 1L
Specific Gravity 1.23
Refractive Index 1.6410 to 1.644
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown to Brown-Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Fieser 15,20
Merck Index 15, 1110
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol and carbon disulfide
Formula Weight 135.18
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Benzothiazole
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning