Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzothiazole, 97%, Thermo Scientific Chemicals

Catalog Number AC146390050
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
95-16-9
C7H5NS
135.18
MFCD00005775
IOJUPLGTWVMSFF-UHFFFAOYSA-N
benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
7222
CHEBI:45993
1,3-benzothiazole
C1=CC=C2C(=C1)N=CS2
95-16-9
C7H5NS
135.18
MFCD00005775
IOJUPLGTWVMSFF-UHFFFAOYSA-N
benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
7222
CHEBI:45993
1,3-benzothiazole
C1=CC=C2C(=C1)N=CS2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 2°C
Color Brown to Brown-Yellow
Density 1.2300g/mL
Boiling Point 231°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6410 to 1.644
Quantity 5 mL
Fieser 15,20
Specific Gravity 1.23
Merck Index 15, 1110
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol and carbon disulfide
Formula Weight 135.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzothiazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-396-2

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit