Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
56602-33-6
C12H22N6OP·PF6
442.27
MGEVGECQZUIPSV-UHFFFAOYSA-N
bop reagent,1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v,castro's reagent,bop-reagent,benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate,ccris 2602,castros reagent,tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate,benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate,1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate
151348
benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
56602-33-6
C12H22N6OP·PF6
442.27
MGEVGECQZUIPSV-UHFFFAOYSA-N
bop reagent,1h-benzo d 1,2,3 triazol-1-yl oxy tris dimethylamino phosphonium hexafluorophosphate v,castro's reagent,bop-reagent,benzotriazol-1-yloxytris dimethylamino-phosphonium hexafluorophosphate,ccris 2602,castros reagent,tri dimethylamino benzotriazol-1-yloxyphosphonium hexafluorophosphate,benzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate,1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate
151348
benzotriazol-1-yloxy-tris(dimethylamino)phosphanium;hexafluorophosphate
CN(C)[P+](N(C)C)(N(C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
Coupling reagent
Melting Point 136.0°C to 140.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 25g
Assay Percent Range 98% min. (HPLC)
Fieser 06,34; 07,14
Formula Weight 442.27
Physical Form Powder
Percent Purity 98+%
Chemical Name or Material Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep

GHS Signal Word: Danger

EINECSNumber : 260-279-1