Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzoyl chloride, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
2500 g
Description
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
Solubility
Miscible with water, ether, benzene, carbon disulfide and carbon tetra chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases and alcohols.
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
Solubility
Miscible with water, ether, benzene, carbon disulfide and carbon tetra chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases and alcohols.
Specifications
Melting Point | °C to 1°C |
Density | 1.211 |
Boiling Point | 197°C to 199°C |
Flash Point | 69°C (156°F) |
Assay Percent Range | ≥99% |
Refractive Index | 1.5535 |
Quantity | 250 g |
UN Number | UN1736 |
Sensitivity | Moisture sensitive |
Merck Index | 14,1112 |
Show More |
Safety and Handling
Hazard Category | H227-H302-H311+H331-H314-H317-H335 |
Hazard Statement | GHS H Statement H314-H302-H312-H332-H317-H227 |
Precautionary Statement | P210-P235-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BENZOYL CHLORIDE |
EINECSNumber | 202-710-8 |
TSCA | Yes |
storageNote1 | Moisture sensitive |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction