Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl Cyanide 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
613-90-1
C8H5NO
131.134
MFCD00001837
GJQBHOAJJGIPRH-UHFFFAOYSA-N
phenylglyoxylonitrile,benzoyl nitrile,benzoylcyanide,oxo phenyl acetonitrile,alpha-oxobenzeneacetonitrile,benzeneacetonitrile, .alpha.-oxo,glyoxylonitrile, phenyl,alpha-oxo-alpha-tolunitrile,2-oxo-2-phenylethanenitrile,alpha-tolunitrile, alpha-oxo
11953
CHEBI:51853
benzoyl cyanide
C1=CC=C(C=C1)C(=O)C#N
613-90-1
C8H5NO
131.134
MFCD00001837
GJQBHOAJJGIPRH-UHFFFAOYSA-N
phenylglyoxylonitrile,benzoyl nitrile,benzoylcyanide,oxo phenyl acetonitrile,alpha-oxobenzeneacetonitrile,benzeneacetonitrile, .alpha.-oxo,glyoxylonitrile, phenyl,alpha-oxo-alpha-tolunitrile,2-oxo-2-phenylethanenitrile,alpha-tolunitrile, alpha-oxo
11953
CHEBI:51853
benzoyl cyanide
C1=CC=C(C=C1)C(=O)C#N
Melting Point 27°C to 32°C
Packaging Glass bottle
Quantity 25g
Color Yellow to Brown
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula C6H5COCN
Formula Weight 131.13
Physical Form Low Melting Mass
Percent Purity ≥94%
Chemical Name or Material Benzoyl cyanide
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger