Alfa Aesar™ Benzoylcholine iodide, 98+%

Manufacturer: Alfa Aesar™ B2238714

Quantity 25g

 View more versions of this product

Catalog No. AAB2238714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17518-43-3
    Molecular Formula C12H18INO2
    Linear Formula C6H5CO2CH2CH2N(CH3)3I
    Formula Weight 335.19
    MDL Number MFCD00038726
    Beilstein 3576708
    Synonym (2-Benzoyloxyethyl)trimethylammonium iodide
    Chemical Name or Material Benzoylcholine iodide

    Specifications

    Formula Weight 335.19
    Molecular Formula C12H18INO2
    Linear Formula C6H5CO2CH2CH2N(CH3)3I
    MDL Number MFCD00038726
    CAS 17518-43-3
    Beilstein 3576708
    Synonym (2-Benzoyloxyethyl)trimethylammonium iodide
    Quantity 25g
    Recommended Storage 2° to 8°C
    TSCA TSCA
    Melting Point ≈245°C (dec.)
    Chemical Name or Material Benzoylcholine iodide
    Assay ≥98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit