Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzoylcholine iodide, 98+%

Click to view available options
Quantity:
1g
5g
25g
17518-43-3
C12H18INO2
335.185
MFCD00038726
XILQCEGWGBSDSH-UHFFFAOYSA-M
benzoylcholine iodide,benzoylcholineiodide,2-benzoyloxy-n,n,n-trimethylethanaminium iodide,2-benzoyloxyethyl trimethylammonium iodide,ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide,ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide 1:1,2-benzoyloxy ethyl trimethylazanium iodide,choline, iodide benzoate,choline, benzoate,acmc-1buu4
87143
2-benzoyloxyethyl(trimethyl)azanium;iodide
C[N+](C)(C)CCOC(=O)C1=CC=CC=C1.[I-]
17518-43-3
C12H18INO2
335.185
MFCD00038726
XILQCEGWGBSDSH-UHFFFAOYSA-M
benzoylcholine iodide,benzoylcholineiodide,2-benzoyloxy-n,n,n-trimethylethanaminium iodide,2-benzoyloxyethyl trimethylammonium iodide,ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide,ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide 1:1,2-benzoyloxy ethyl trimethylazanium iodide,choline, iodide benzoate,choline, benzoate,acmc-1buu4
87143
2-benzoyloxyethyl(trimethyl)azanium;iodide
C[N+](C)(C)CCOC(=O)C1=CC=CC=C1.[I-]
Quantity 5g
Melting Point ~245°C (decomposition)
Linear Formula C6H5CO2CH2CH2N(CH3)3I
Beilstein 3576708
Formula Weight 335.19
Percent Purity ≥98%
Chemical Name or Material Benzoylcholine iodide

TSCA : TSCA

Recommended Storage : 2° to 8°C