Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benztropine Mesylate, USP, 98-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
Packaging:
Amber Glass Bottle
132-17-2
C22H29NO4S
403.54
CPFJLLXFNPCTDW-STYNFMPRSA-N
(1R,5R)-3-(diphenylmethoxy)-8-methyl-8-azabicyclo[3.2.1]octane; methanesulfonic acid
CS(O)(=O)=O.CN1[C@@H]2CC[C@@H]1CC(C2)OC(C1=CC=CC=C1)C1=CC=CC=C1
132-17-2
C22H29NO4S
403.54
CPFJLLXFNPCTDW-STYNFMPRSA-N
(1R,5R)-3-(diphenylmethoxy)-8-methyl-8-azabicyclo[3.2.1]octane; methanesulfonic acid
CS(O)(=O)=O.CN1[C@@H]2CC[C@@H]1CC(C2)OC(C1=CC=CC=C1)C1=CC=CC=C1

B1307, 132-17-2, C21H25NO•CH4O3S

Spectrum™ Benztropine Mesylate, USP is an anticholinergic agent. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Melting Range 141-148°C
Loss on Drying 0.05
Quantity 1 kg
CAS Max % 1
Percent Purity 98 to 100.5%