Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 2-bromoethyl ether, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
100g
1462-37-9
C9H11BrO
215.09
MFCD01321307
FWOHDAGPWDEWIB-UHFFFAOYSA-N
benzyl 2-bromoethyl ether,2-bromoethoxy methyl benzene,2-benzyloxy-1-bromoethane,benzyloxyethyl bromide,benzene, 2-bromoethoxy methyl,1-bromo-2-benzyloxyethane,2-bromoethoxymethyl benzene,2-bromo-ethoxymethyl-benzene
73833
2-bromoethoxymethylbenzene
C1=CC=C(C=C1)COCCBr
1462-37-9
C9H11BrO
215.09
MFCD01321307
FWOHDAGPWDEWIB-UHFFFAOYSA-N
benzyl 2-bromoethyl ether,2-bromoethoxy methyl benzene,2-benzyloxy-1-bromoethane,benzyloxyethyl bromide,benzene, 2-bromoethoxy methyl,1-bromo-2-benzyloxyethane,2-bromoethoxymethyl benzene,2-bromo-ethoxymethyl-benzene
73833
2-bromoethoxymethylbenzene
C1=CC=C(C=C1)COCCBr
Specific Gravity 1.36
Quantity 25g
Packaging Glass bottle
Boiling Point 254.0°C
Density 1.3600g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5CH2OCH2CH2Br
Formula Weight 215.09
Percent Purity 97%
Chemical Name or Material Benzyl 2-bromoethyl ether
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning