Alfa Aesar™ Benzyl 2-naphthyl ether, 99%

Manufacturer: Alfa Aesar™ B2179636

Quantity 500g

 View more versions of this product

Catalog No. AAB2179636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 613-62-7
    Molecular Formula C17H14O
    Formula Weight 234.3
    MDL Number MFCD00089092
    Beilstein 2049922
    Synonym 2-Benzyloxynaphthalene
    Chemical Name or Material Benzyl 2-naphthyl ether

    Specifications

    Formula Weight 234.3
    Molecular Formula C17H14O
    MDL Number MFCD00089092
    CAS 613-62-7
    Beilstein 2049922
    Synonym 2-Benzyloxynaphthalene
    Melting Point 96° to 98°C
    Quantity 500g
    Chemical Name or Material Benzyl 2-naphthyl ether
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit