Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl 3-bromopropyl ether, 95%

Click to view available options
Quantity:
1g
5g
54314-84-0
C10H13BrO
229.117
MFCD00134570
PSUXTZLDBVEZTD-UHFFFAOYSA-N
benzyl 3-bromopropyl ether,3-bromopropoxy methyl benzene,3-bromopropoxy methyl benzene,3-bromopropylbenzyl ether,3-benzyloxy-1-bromopropane,3-benzyloxy propyl bromide,benzene, 3-bromopropoxy methyl,1-3-bromopropoxy methyl benzene,1-3-bromopropoxy methyl benzene,benzyl3-bromopropylether
2776064
3-bromopropoxymethylbenzene
C1=CC=C(C=C1)COCCCBr
54314-84-0
C10H13BrO
229.117
MFCD00134570
PSUXTZLDBVEZTD-UHFFFAOYSA-N
benzyl 3-bromopropyl ether,3-bromopropoxy methyl benzene,3-bromopropoxy methyl benzene,3-bromopropylbenzyl ether,3-benzyloxy-1-bromopropane,3-benzyloxy propyl bromide,benzene, 3-bromopropoxy methyl,1-3-bromopropoxy methyl benzene,1-3-bromopropoxy methyl benzene,benzyl3-bromopropylether
2776064
3-bromopropoxymethylbenzene
C1=CC=C(C=C1)COCCCBr
Applications
Benzyl 3-bromopropyl ether may be used in the studies of preparation of (2S,3S)-1-[(triisopropylsilyl)oxy]-7-(benzyloxy)-2,3-(isopropylidenedioxy)-4(Z)-heptene, Ppeparation of 5-(3-Benzyloxypropoxy)psoralen (PAP-7), total synthesis of zincophorin and (+)-anatoxin-a.

Solubility
Slightly soluble in water.

Notes
Store under dry inert gas. Store away from air. Incompatible with air and oxidizing agents.
Color Colorless
Refractive Index 1.531
Quantity 5g
Density 1.298g/mL
Boiling Point 130°C to 132°C (8mmHg)
Beilstein 2084412
Formula Weight 229.12
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Benzyl 3-bromopropyl ether

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning