Learn More
Benzyl 3-bromopropyl ether, 95%, Thermo Scientific Chemicals
Description
Benzyl 3-bromopropyl ether may be used in the studies of preparation of (2S,3S)-1-[(triisopropylsilyl)oxy]-7-(benzyloxy)-2,3-(isopropylidenedioxy)-4(Z)-heptene, Ppeparation of 5-(3-Benzyloxypropoxy)psoralen (PAP-7), total synthesis of zincophorin and (+)-anatoxin-a.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzyl 3-bromopropyl ether may be used in the studies of preparation of (2S,3S)-1-[(triisopropylsilyl)oxy]-7-(benzyloxy)-2,3-(isopropylidenedioxy)-4(Z)-heptene, Ppeparation of 5-(3-Benzyloxypropoxy)psoralen (PAP-7), total synthesis of zincophorin and (+)-anatoxin-a.
Solubility
Slightly soluble in water.
Notes
Store under dry inert gas. Store away from air. Incompatible with air and oxidizing agents.
Specifications
Color | Colorless |
Density | 1.298 g/mL |
Boiling Point | 130°C to 132°C (8 mmHg) |
Flash Point | 113°C (235°F) |
Refractive Index | 1.531 |
Quantity | 1 g |
Beilstein | 2084412 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 229.12 |
Percent Purity | 95% |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.