Alfa Aesar™ Benzyl 3-hydroxyphenylacetate, 98%

Manufacturer: Alfa Aesar™ B2467903

Quantity 1g

 View more versions of this product

Catalog No. AAB2467903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 295320-25-1
    Molecular Formula C15H14O3
    Formula Weight 242.27
    MDL Number MFCD04039768
    Synonym 3-Hydroxyphenylacetic acid benzyl ester
    Chemical Name or Material Benzyl 3-hydroxyphenylacetate

    Specifications

    Formula Weight 242.27
    MDL Number MFCD04039768
    Quantity 1g
    Molecular Formula C15H14O3
    CAS 295320-25-1
    Synonym 3-Hydroxyphenylacetic acid benzyl ester
    Melting Point 79° to 81°C
    Chemical Name or Material Benzyl 3-hydroxyphenylacetate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit