Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl 3-hydroxyphenylacetate, 98%

Click to view available options
Quantity:
1g
5g
25g
295320-25-1
C15H14O3
242.274
MFCD04039768
ALFOBMRIXXPJLQ-UHFFFAOYSA-N
benzyl 3-hydroxyphenylacetate,benzyl 2-3-hydroxyphenyl acetate,acmc-20alpo,benzyl-3-hydroxyphenylacetate,3-hydroxyphenylacetic acid benzyl ester,phenylmethyl 2-3-hydroxyphenyl acetate,3-hydroxy-phenyl-acetic acid benzyl ester
7016416
benzyl 2-(3-hydroxyphenyl)acetate
C1=CC=C(C=C1)COC(=O)CC2=CC(=CC=C2)O
295320-25-1
C15H14O3
242.274
MFCD04039768
ALFOBMRIXXPJLQ-UHFFFAOYSA-N
benzyl 3-hydroxyphenylacetate,benzyl 2-3-hydroxyphenyl acetate,acmc-20alpo,benzyl-3-hydroxyphenylacetate,3-hydroxyphenylacetic acid benzyl ester,phenylmethyl 2-3-hydroxyphenyl acetate,3-hydroxy-phenyl-acetic acid benzyl ester
7016416
benzyl 2-(3-hydroxyphenyl)acetate
C1=CC=C(C=C1)COC(=O)CC2=CC(=CC=C2)O
Melting Point 79°C to 81°C
Quantity 5g
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material Benzyl 3-hydroxyphenylacetate