Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 3-pyrroline-1-carboxylate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
31970-04-4
C12H13NO2
203.24
XSKKIFJNZPNVGO-UHFFFAOYSA-N
benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate,benzyl 3-pyrroline-1-carboxylate,1-cbz-2,5-dihydro-1h-pyrrole,n-cbz-2,5-dihydropyrrole,n-cbz-3-pyrroline,n-carbobenzoxy-3-pyrroline,2,5-dihydropyrrole-1-carboxylic acid benzyl ester,benzyl 2h-pyrrole-1 5h-carboxylate,n-benzyloxycarbonyl-3-pyrroline,1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
643471
benzyl 2,5-dihydropyrrole-1-carboxylate
C1C=CCN1C(=O)OCC2=CC=CC=C2
31970-04-4
C12H13NO2
203.24
XSKKIFJNZPNVGO-UHFFFAOYSA-N
benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate,benzyl 3-pyrroline-1-carboxylate,1-cbz-2,5-dihydro-1h-pyrrole,n-cbz-2,5-dihydropyrrole,n-cbz-3-pyrroline,n-carbobenzoxy-3-pyrroline,2,5-dihydropyrrole-1-carboxylic acid benzyl ester,benzyl 2h-pyrrole-1 5h-carboxylate,n-benzyloxycarbonyl-3-pyrroline,1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
643471
benzyl 2,5-dihydropyrrole-1-carboxylate
C1C=CCN1C(=O)OCC2=CC=CC=C2
Color Colorless to Yellow
Specific Gravity 1.132
Infrared Spectrum Authentic
Density 1.1320g/mL
Flash Point >110°C
Boiling Point 133.0°C to 137.0°C (0.6mmHg)
Packaging Glass bottle
Refractive Index 1.544
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 203.24
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Benzyl 3-pyrroline-1-carboxylate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and k

GHS Signal Word: Warning