Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl 4-aminopiperidine-1-carboxylate, 97%

Click to view available options
Quantity:
250mg
1g
5g
120278-07-1
C13H18N2O2
234.299
MFCD05863884
YYIQGSYCCNQAGV-UHFFFAOYSA-N
1-n-cbz-4-aminopiperidine,4-amino-1-n-cbz-piperidine,1-cbz-4-aminopiperidine,4-amino-piperidine-1-carboxylic acid benzyl ester,4-amino-1-cbz-piperidine,1-n-cbz-4-amino-piperidine,1-piperidinecarboxylic acid, 4-amino-, phenylmethyl ester,4-aminopiperidine-1-carboxylic acid benzyl ester,benzyl 4-aminopiperidine-1-carboxylate; 4-aminopiperidine-1-carboxylic acid benzyl ester,pubchem19775
17846640
benzyl 4-aminopiperidine-1-carboxylate
C1CN(CCC1N)C(=O)OCC2=CC=CC=C2
120278-07-1
C13H18N2O2
234.299
MFCD05863884
YYIQGSYCCNQAGV-UHFFFAOYSA-N
1-n-cbz-4-aminopiperidine,4-amino-1-n-cbz-piperidine,1-cbz-4-aminopiperidine,4-amino-piperidine-1-carboxylic acid benzyl ester,4-amino-1-cbz-piperidine,1-n-cbz-4-amino-piperidine,1-piperidinecarboxylic acid, 4-amino-, phenylmethyl ester,4-aminopiperidine-1-carboxylic acid benzyl ester,benzyl 4-aminopiperidine-1-carboxylate; 4-aminopiperidine-1-carboxylic acid benzyl ester,pubchem19775
17846640
benzyl 4-aminopiperidine-1-carboxylate
C1CN(CCC1N)C(=O)OCC2=CC=CC=C2
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 1g
Beilstein 10003356
Formula Weight 234.3
Percent Purity 97%
Chemical Name or Material Benzyl 4-aminopiperidine-1-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning