Alfa Aesar™ Benzyl 4-bromophenyl ketone, 98%

Manufacturer: Alfa Aesar™ B2215903

Quantity 1g

 View more versions of this product

Catalog No. AAB2215903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2001-29-8
    Molecular Formula C14H11BrO
    Formula Weight 275.14
    MDL Number MFCD00016331
    Beilstein 2211322
    Synonym 4-Bromodeoxybenzoin; 4'-Bromo-2-phenylacetophenone
    Chemical Name or Material Benzyl 4-bromophenyl ketone

    Specifications

    Formula Weight 275.14
    Melting Point 109° to 112°C
    Quantity 1g
    Molecular Formula C14H11BrO
    MDL Number MFCD00016331
    CAS 2001-29-8
    Beilstein 2211322
    Synonym 4-Bromodeoxybenzoin; 4'-Bromo-2-phenylacetophenone
    Chemical Name or Material Benzyl 4-bromophenyl ketone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit